N-BOC-L-tert-Leucine, 98%, Thermo Scientific Chemicals
N-BOC-L-tert-Leucine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-BOC-L-tert-Leucine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass Bottle
Catalog number 368010250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS62965-35-9
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid
Molecular FormulaC11H21NO4
InChI KeyLRFZIPCTFBPFLX-JLDDOWRYNA-N
SMILESCC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to light yellow
Titration with NaOH>=97.5 %
Specific optical rotation-3.0° to -5.0° (20°C, 589 nm) (c=1, acetic acid)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only