Thermo Scientific Chemicals

3-Amino-5-methylpyrazole, 97%, Thermo Scientific Chemicals

Catalog number: 368031000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Amino-5-methylpyrazole, 97%, Thermo Scientific Chemicals

Catalog number: 368031000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 368031000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
31230-17-8
IUPAC Name
5-methyl-1H-pyrazol-3-amine
Molecular Formula
C4H7N3
InChI Key
FYTLHYRDGXRYEY-UHFFFAOYSA-N
SMILES
CC1=CC(N)=NN1
Appearance (Color)
Light yellow to orange
Appearance (Form)
Solid or viscous liquid
GC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text