Zinc cyanide, 98%, pure, Thermo Scientific Chemicals
Zinc cyanide, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc cyanide, 98%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
250 g
Packaging:
View all
Plastic bottle
Catalog number 368070010
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
Add to cart
Zinc cyanide, 98%, pure, Thermo Scientific Chemicals
Catalog number368070010
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS557-21-1
IUPAC Namezinc cyanide
Molecular FormulaC2N2Zn
InChI KeyGTLDTDOJJJZVBW-UHFFFAOYSA-N
SMILES[Zn++].[C-]#N.[C-]#N
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=97.5 %
Appearance (Color)White to off-white
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only