Thermo Scientific Chemicals

Stearyl stearate, 98%, Thermo Scientific Chemicals

Catalog number: 368110010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Stearyl stearate, 98%, Thermo Scientific Chemicals

Catalog number: 368110010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 368110010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2778-96-3
IUPAC Name
octadecyl octadecanoate
Molecular Formula
C36H72O2
InChI Key
NKBWPOSQERPBFI-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC
Appearance (Form)
Powder or platelets
Infrared spectrum
Conforms
Appearance (Color)
White
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text