1x1 image pixel for data collection
Stearyl stearate, 98%
Stearyl stearate, 98%
Stearyl stearate, 98%
Stearyl stearate, 98%
Thermo Scientific Chemicals

Stearyl stearate, 98%

CAS: 2778-96-3 | C36H72O2 | 536.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 368110050
Price (USD)
531.65
Online Exclusive
591.00
Save 59.35 (10%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
531.65
Online Exclusive
591.00
Save 59.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2778-96-3
IUPAC Nameoctadecyl octadecanoate
Molecular FormulaC36H72O2
InChI KeyNKBWPOSQERPBFI-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or platelets
Infrared spectrumConforms
Appearance (Color)White
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link