3-Bromo-3-buten-1-ol, 97+%, Thermo Scientific Chemicals
3-Bromo-3-buten-1-ol, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-3-buten-1-ol, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 368210500
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS76334-36-6
IUPAC Name3-bromobut-3-en-1-ol
Molecular FormulaC4H7BrO
InChI KeyRTKMFQOHBDVEBC-UHFFFAOYSA-N
SMILESOCCC(Br)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4990 to 1.5020 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow to brown
GC>=97.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only