Thermo Scientific Chemicals

Methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: 368280050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: 368280050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 368280050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1458-18-0
IUPAC Name
methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
Molecular Formula
C6H5Cl2N3O2
InChI Key
USYMCUGEGUFUBI-UHFFFAOYSA-N
SMILES
COC(=O)C1=NC(Cl)=C(Cl)N=C1N
Infrared spectrum
Conforms
Melting point
229°C to 235°C (Dec)
Appearance (Color)
Light brown to red-brown
Appearance (Form)
Powder
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text