Thermo Scientific Chemicals

4-Bromopiperidine hydrobromide, 98%, Thermo Scientific Chemicals

Catalog number: 368310010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromopiperidine hydrobromide, 98%, Thermo Scientific Chemicals

Catalog number: 368310010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 368310010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54288-70-9
IUPAC Name
4-bromopiperidin-1-ium
Molecular Formula
C5H11BrN
InChI Key
FIHYVUSUEHIGOM-UHFFFAOYSA-O
SMILES
BrC1CC[NH2+]CC1
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration Argentometric
97.5 to 102.5 %
Appearance (Color)
White to beige
Melting point
191°C to 196°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text