Thermo Scientific Chemicals

(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, Thermo Scientific Chemicals

Catalog number: 368321000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, Thermo Scientific Chemicals

Catalog number: 368321000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 368321000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
81290-20-2
IUPAC Name
tert-butyl(trifluoromethyl)silane
Molecular Formula
C5H11F3Si
InChI Key
MTYSDPUZDZURPP-UHFFFAOYSA-N
SMILES
CC(C)(C)[SiH2]C(F)(F)F
Appearance (Color)
Clear colorless
Strength
0.45 to 0.55 M
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text