Pyrrole-2-carbonitrile, 97%, Thermo Scientific Chemicals
Pyrrole-2-carbonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyrrole-2-carbonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 368350050
Price (USD)/ Each
165.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
195.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
165.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
195.00 
Save 29.35 (15%)
Add to cart
Pyrrole-2-carbonitrile, 97%, Thermo Scientific Chemicals
Catalog number368350050
Price (USD)/ Each
165.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
195.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
CAS4513-94-4
IUPAC Name1H-pyrrole-2-carbonitrile
Molecular FormulaC5H4N2
InChI KeyBQMPGKPTOHKYHS-UHFFFAOYSA-N
SMILESN#CC1=CC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear yellow to orange
Infrared spectrumConforms
Refractive index1.5480 to 1.5510 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only