Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™
Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™
Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™
Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™
Thermo Scientific Chemicals

Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™

CAS: 7664-41-7 | H3N | 17.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 368381000
Price (USD)
109.00
Each
In stock
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
109.00
Each
Add to cart
Chemical Identifiers
CAS7664-41-7
IUPAC Nameammonia
Molecular FormulaH3N
InChI KeyQGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILESN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Water=<1 % (K.F.)
Appearance (Color)Clear colorless
Strength0.4 to 0.6 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link