Ammonia, 2M solution in methanol, AcroSeal™, Thermo Scientific Chemicals
Ammonia, 2M solution in methanol, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonia, 2M solution in methanol, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 368398000
Price (USD)/ Each
226.65
Online exclusive
252.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
226.65
Online exclusive
252.00 
Save 25.35 (10%)
Add to cart
Ammonia, 2M solution in methanol, AcroSeal™, Thermo Scientific Chemicals
Catalog number368398000
Price (USD)/ Each
226.65
Online exclusive
252.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7664-41-7, 67-56-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Water=<0.3 %
Appearance (Form)Solution
Strength1.90 to 2.20 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only