(1S,2R)-2-Amino-1,2-diphenylethanol, 98%, Thermo Scientific Chemicals
(1S,2R)-2-Amino-1,2-diphenylethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1S,2R)-2-Amino-1,2-diphenylethanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 368410010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS23364-44-5
IUPAC Name(1R,2S)-2-hydroxy-1,2-diphenylethan-1-aminium
Molecular FormulaC14H16NO
InChI KeyGEJJWYZZKKKSEV-KGLIPLIRSA-O
SMILES[NH3+][C@@H]([C@@H](O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point141°C to 145°C
Appearance (Color)White to beige
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only