Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99.5%, for HPLC, Thermo Scientific Chemicals

Catalog number: 368451000
100 mL, Glass Bottle, Each
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99.5%, for HPLC, Thermo Scientific Chemicals

Catalog number: 368451000
100 mL, Glass Bottle, Each
Quantity
Packaging
Catalog number: 368451000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
872-50-4
IUPAC Name
1-methylpyrrolidin-2-one
Molecular Formula
C5H9NO
InChI Key
SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES
CN1CCCC1=O
GC
>=99.5 %
UV
at 285 nm A: =<1.0
UV
at 300 nm A: =<0.50
UV
at 320 nm A: =<0.10
UV
at 450 nm A: =<0.01

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For HPLC
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text