Thermo Scientific Chemicals

3-Iodobenzonitrile, 97%, Thermo Scientific Chemicals

Catalog number: 368600100
10 g, Each
Thermo Scientific Chemicals

3-Iodobenzonitrile, 97%, Thermo Scientific Chemicals

Catalog number: 368600100
10 g, Each
Quantity
Catalog number: 368600100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69113-59-3
IUPAC Name
3-iodobenzonitrile
Molecular Formula
C7H4IN
InChI Key
BGARPMGQRREXLN-UHFFFAOYSA-N
SMILES
IC1=CC=CC(=C1)C#N
Appearance (Color)
White to brown
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
38°C to 44°C
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text