Thermo Scientific Chemicals

Phosphomolybdic acid, 20 wt.% solution in ethanol, Thermo Scientific Chemicals

Catalog number: 368621000
100 mL, Each
Thermo Scientific Chemicals

Phosphomolybdic acid, 20 wt.% solution in ethanol, Thermo Scientific Chemicals

Catalog number: 368621000
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 368621000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64-17-5
Titration with NaOH
19 to 21 %
Appearance (Color)
Clear yellow to green
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text