Thermo Scientific Chemicals

Methyl 3-(bromomethyl)benzoate, 95%, Thermo Scientific Chemicals

Catalog number: 368630050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3-(bromomethyl)benzoate, 95%, Thermo Scientific Chemicals

Catalog number: 368630050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 368630050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1129-28-8
IUPAC Name
methyl 3-(bromomethyl)benzoate
Molecular Formula
C9H9BrO2
InChI Key
YUHSMQQNPRLEEJ-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(CBr)=C1
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
GC
>=94 %
Melting point
41°C to 47°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text