Thermo Scientific Chemicals

(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%, Thermo Scientific Chemicals

Catalog number: 368641000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%, Thermo Scientific Chemicals

Catalog number: 368641000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 368641000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
98327-87-8
IUPAC Name
[2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane
Molecular Formula
C44H32P2
InChI Key
MUALRAIOVNYAIW-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=C(C2=CC=CC=C2C=C1)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1
HPLC
>=97.5 %
Melting point
280°C to 285°C
Appearance (Color)
White to light beige
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text