Thermo Scientific Chemicals

(Triethylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Catalog number: 368730050
Each
Thermo Scientific Chemicals

(Triethylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Catalog number: 368730050
Each
Catalog number: 368730050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1777-03-3
IUPAC Name
triethyl(ethynyl)silane
Molecular Formula
C8H16Si
InChI Key
FWSPXZXVNVQHIF-UHFFFAOYSA-N
SMILES
CC[Si](CC)(CC)C#C
Refractive index
1.4310 to 1.4330 (20°C, 589 nm)
GC
>=96.0 %
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text