Thermo Scientific Chemicals

(Triisopropylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Catalog number: 368740050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

(Triisopropylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Catalog number: 368740050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 368740050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89343-06-6
IUPAC Name
ethynyltris(propan-2-yl)silane
Molecular Formula
C11H22Si
InChI Key
KZGWPHUWNWRTEP-UHFFFAOYSA-N
SMILES
CC(C)[Si](C#C)(C(C)C)C(C)C
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.4515 to 1.4535 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text