Thermo Scientific Chemicals

Triethyl 2-phosphonobutyrate, 98%, Thermo Scientific Chemicals

Catalog number: 368780250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Triethyl 2-phosphonobutyrate, 98%, Thermo Scientific Chemicals

Catalog number: 368780250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 368780250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17145-91-4
IUPAC Name
ethyl 2-(diethoxyphosphoryl)butanoate
Molecular Formula
C10H21O5P
InChI Key
GYUCVQSNZFRDRL-UHFFFAOYNA-N
SMILES
CCOC(=O)C(CC)P(=O)(OCC)OCC
Refractive index
1.4315 to 1.4335 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text