2-Aminoimidazole hemisulfate, 98+%, Thermo Scientific Chemicals
2-Aminoimidazole hemisulfate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminoimidazole hemisulfate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Catalog number 368860025
Price (USD)/ Each
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1450-93-7
IUPAC Namebis(1H-imidazol-2-amine); sulfuric acid
Molecular FormulaC6H12N6O4S
InChI KeyKUWRLKJYNASPQZ-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NC1=NC=CN1.NC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=98.0 %
Appearance (Color)Brown
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only