Thermo Scientific Chemicals

2-Aminoimidazole hemisulfate, 98+%, Thermo Scientific Chemicals

Catalog number: 368860100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Aminoimidazole hemisulfate, 98+%, Thermo Scientific Chemicals

Catalog number: 368860100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 368860100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1450-93-7
IUPAC Name
bis(1H-imidazol-2-amine); sulfuric acid
Molecular Formula
C6H12N6O4S
InChI Key
KUWRLKJYNASPQZ-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.NC1=NC=CN1.NC1=NC=CN1
Infrared spectrum
Conforms
Titration with NaOH
>=98.0 %
Appearance (Color)
Brown
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text