[Bis(2-methoxyethyl)amino]sulfur trifluoride, 2.7M (50 wt.%) solution in toluene, Thermo Scientific Chemicals
[Bis(2-methoxyethyl)amino]sulfur trifluoride, 2.7M (50 wt.%) solution in toluene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

[Bis(2-methoxyethyl)amino]sulfur trifluoride, 2.7M (50 wt.%) solution in toluene, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 368961000
Price (USD)/ Each
530.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
624.00 
Save 93.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
530.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
624.00 
Save 93.35 (15%)
Add to cart
[Bis(2-methoxyethyl)amino]sulfur trifluoride, 2.7M (50 wt.%) solution in toluene, Thermo Scientific Chemicals
Catalog number368961000
Price (USD)/ Each
530.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
624.00 
Save 93.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namebis(2-methoxyethyl)(trifluoro-λ⁴-sulfanyl)amine
Molecular FormulaC6H14F3NO2S
InChI KeyAPOYTRAZFJURPB-UHFFFAOYSA-N
SMILESCOCCN(CCOC)S(F)(F)F
Molecular Weight (g/mol)221.24
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO448 to 55 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only