Thermo Scientific Chemicals

tert-Butylchlorodimethylsilane, 1.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 369101000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

tert-Butylchlorodimethylsilane, 1.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 369101000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 369101000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18162-48-6
IUPAC Name
tert-butyl(chloro)dimethylsilane
Molecular Formula
C6H15ClSi
InChI Key
BCNZYOJHNLTNEZ-UHFFFAOYSA-N
SMILES
CC(C)(C)[Si](C)(C)Cl
Appearance (Color)
Colorless
Appearance (Form)
Solution
Infrared spectrum
Conforms
Titration Argentometric
0.95 to 1.10 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text