Thermo Scientific Chemicals

N-(Diphenylmethylene)glycine tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: 369320010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(Diphenylmethylene)glycine tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: 369320010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 369320010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
81477-94-3
IUPAC Name
tert-butyl 2-[(diphenylmethylidene)amino]acetate
Molecular Formula
C19H21NO2
InChI Key
YSHDPXQDVKNPKA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
Melting point
111°C to 115°C
Appearance (Form)
Crystalline powder
Appearance (Color)
White
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text