Thermo Scientific Chemicals

tert-Butyl 2,2,2-trichloroacetimidate, 95%, Thermo Scientific Chemicals

Catalog number: 369330250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

tert-Butyl 2,2,2-trichloroacetimidate, 95%, Thermo Scientific Chemicals

Catalog number: 369330250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 369330250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
98946-18-0
IUPAC Name
tert-butyl 2,2,2-trichloroethanimidate
Molecular Formula
C6H10Cl3NO
InChI Key
CQXDYHPBXDZWBA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=N)C(Cl)(Cl)Cl
Appearance (Form)
Liquid or low melting crystalline mass
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4550 to 1.4570 (20°C, 589 nm)
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text