3,4-Dimethoxyphenylboronic acid, 97%, Thermo Scientific Chemicals
3,4-Dimethoxyphenylboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dimethoxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 369390050
Price (USD)/ Each
69.60
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
69.60
Add to cart
3,4-Dimethoxyphenylboronic acid, 97%, Thermo Scientific Chemicals
Catalog number369390050
Price (USD)/ Each
69.60
-
Add to cart
Chemical Identifiers
CAS122775-35-3
IUPAC Name(3,4-dimethoxyphenyl)boronic acid
Molecular FormulaC8H11BO4
InChI KeyRCVDPBFUMYUKPB-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1OC)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or granules and/or chunks
Appearance (Color)White to beige
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only