Thermo Scientific Chemicals

4-Acetylphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 369400250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Acetylphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 369400250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 369400250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
149104-90-5
IUPAC Name
(4-acetylphenyl)boronic acid
Molecular Formula
C8H9BO3
InChI Key
OBQRODBYVNIZJU-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(C=C1)B(O)O
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=96.0 %
Appearance (Color)
White to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text