1,11-Undecanedicarboxylic acid, Thermo Scientific Chemicals
1,11-Undecanedicarboxylic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,11-Undecanedicarboxylic acid, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 369440250
Price (USD)/ Each
116.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
116.00
Add to cart
1,11-Undecanedicarboxylic acid, Thermo Scientific Chemicals
Catalog number369440250
Price (USD)/ Each
116.00
-
Add to cart
Chemical Identifiers
CAS505-52-2
IUPAC Nametridecanedioic acid
Molecular FormulaC13H24O4
InChI KeyDXNCZXXFRKPEPY-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
GC>=92 %
Infrared spectrumConforms
Melting point112°C to 116°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only