1-Decanesulfonic acid, sodium salt, 96%, Thermo Scientific Chemicals
1-Decanesulfonic acid, sodium salt, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Decanesulfonic acid, sodium salt, 96%, Thermo Scientific Chemicals

1-Decanesulfonic acid, sodium salt, 96%, ACROS Organics, Quantity: 25g, Packaging: Glass bottle, White to Cream, Melting Point: >300 deg.C, Molecular Weight: 244.32, Percent Purity: 96%, Assay Percent Range: 96%, Beilstein: 04, III, 27, CAS: 13419-61-9, CAS Max %: 100.0, CAS Min %: 95.0
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 369450250
Price (USD)/ Each
194.65
Online exclusive
216.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
194.65
Online exclusive
216.00 
Save 21.35 (10%)
Add to cart
1-Decanesulfonic acid, sodium salt, 96%, Thermo Scientific Chemicals
Catalog number369450250
Price (USD)/ Each
194.65
Online exclusive
216.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13419-61-9
IUPAC Namesodium decane-1-sulfonate
Molecular FormulaC10H21NaO3S
InChI KeyAIMUHNZKNFEZSN-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<5 % (100°C, 3 h) (vacuum)
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
Titration after Ion exchange>=95 % (On dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only