6-Aminoquinoline, 98%, Thermo Scientific Chemicals
6-Aminoquinoline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Aminoquinoline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 369640050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS580-15-4
IUPAC Namequinolin-6-amine
Molecular FormulaC9H8N2
InChI KeyRJSRSRITMWVIQT-UHFFFAOYSA-N
SMILESNC1=CC=C2N=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO497.5 to 102.5 %
Appearance (Form)Powder
Melting point115°C to 119°C
Appearance (Color)Yellow to grey-green to brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only