Search Thermo Fisher Scientific
Catalog Number | Description | Plug Type |
---|---|---|
369644 | Electric Screwdriver, European Plug | European |
369643 | Manual Screwdriver | US |
Quickly and easily cap and decap Nunc Cryobank tubes using Thermo Scientific™ Nunc™ Screwdrivers for Cryobank and Bank-It™ Systems, available in manual and electric models to suit your application. Nunc Cryobank vials provide the customer with superior tracking of their cryovials using a 2D code, the highest storage density on the market and the ability to automate the capping and decapping of the vials. The tubes click into the rack and can be handled without the risk of accidentally dropping a vial.