Thermo Scientific Chemicals

Di-tert-butyl iminodicarboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 369660250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

Di-tert-butyl iminodicarboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 369660250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 369660250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
51779-32-9
IUPAC Name
tert-butyl N-[(tert-butoxy)carbonyl]carbamate
Molecular Formula
C10H19NO4
InChI Key
XCAQIUOFDMREBA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NC(=O)OC(C)(C)C
Appearance (Form)
Crystalline powder
Appearance (Color)
White to beige
GC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text