Thermo Scientific Chemicals

Palladium(II) chloride, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 369670250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Palladium(II) chloride, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 369670250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 369670250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7647-10-1
IUPAC Name
palladium(2+) dichloride
Molecular Formula
Cl2Pd
InChI Key
PIBWKRNGBLPSSY-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Pd++]
Appearance (Color)
Brown
Total trace metal impurities
=<0.001 %
Appearance (Form)
Powder
Assay
59 to 61 % (Pd)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text