Thermo Scientific Chemicals

Cerium(III) chloride, 99.9%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 369751000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Cerium(III) chloride, 99.9%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 369751000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 369751000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-86-5
IUPAC Name
cerium(3+) trichloride
Molecular Formula
CeCl3
InChI Key
VYLVYHXQOHJDJL-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ce+3]
Appearance (Color)
White to light yellow
Appearance (Form)
Powder or crystals or granules
Total conc. of imp.as REO
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text