(S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, Thermo Scientific Chemicals
(S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number 369770250
Price (USD)/ Each
222.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
262.00 
Save 39.35 (15%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
222.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
262.00 
Save 39.35 (15%)
Add to cart
(S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, Thermo Scientific Chemicals
Catalog number369770250
Price (USD)/ Each
222.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
262.00 
Save 39.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(3aS)-1-methyl-3,3-diphenyl-hexahydropyrrolo[1,2-c][1,3,2]oxazaborole
Molecular FormulaC18H20BNO
InChI KeyVMKAFJQFKBASMU-KRWDZBQOSA-N
SMILES[H][C@@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)277.17
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Colorless to amber
Strength0.95 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only