Thermo Scientific Chemicals

Isosorbide dimethyl ether, 99%, Thermo Scientific Chemicals

Catalog number: 369910250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Isosorbide dimethyl ether, 99%, Thermo Scientific Chemicals

Catalog number: 369910250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 369910250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5306-85-4
IUPAC Name
3,6-dimethoxy-hexahydrofuro[3,2-b]furan
Molecular Formula
C8H14O4
InChI Key
MEJYDZQQVZJMPP-UHFFFAOYNA-N
SMILES
COC1COC2C(COC12)OC
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4600 to 1.4620 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text