Isosorbide dimethyl ether, 99%
Isosorbide dimethyl ether, 99%
Isosorbide dimethyl ether, 99%
Isosorbide dimethyl ether, 99%
Thermo Scientific Chemicals

Isosorbide dimethyl ether, 99%

CAS: 5306-85-4 | C8H14O4 | 174.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 369911000
Price (USD)
182.65
Special offer
Online exclusive
215.00
Save 32.35 (15%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
182.65
Special offer
Online exclusive
215.00
Save 32.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS5306-85-4
IUPAC Name3,6-dimethoxy-hexahydrofuro[3,2-b]furan
Molecular FormulaC8H14O4
InChI KeyMEJYDZQQVZJMPP-UHFFFAOYNA-N
SMILESCOC1COC2C(COC12)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.4600 to 1.4620 (20°C, 589 nm)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470488Certificate of AnalysisMay 17, 2025369910250, 369911000
A0458091Certificate of AnalysisNov 29, 2023369910250, 369911000
A0425578Certificate of AnalysisAug 25, 2023369910250, 369911000
A0448291Certificate of AnalysisDec 15, 2022369910250, 369911000
A0399767Certificate of AnalysisDec 14, 2022369910250, 369911000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection