Thermo Scientific Chemicals

Amberlite™ IRA-402, Cl-form, ion-exchange resin, Thermo Scientific Chemicals

Catalog number: 369940050
5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Amberlite™ IRA-402, Cl-form, ion-exchange resin, Thermo Scientific Chemicals

Catalog number: 369940050
5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 369940050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
52439-77-7
MDL Number
MFCD00145564
Appearance (Color)
Light yellow
Total volume capacity
>=1.20 eq/l (Cl)
Harmonic mean size
0.600 to 0.750 mm
Moisture holding capacity
49.0 to 60.0 % (Cl)
Uniformity coefficient
=<1.60

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text