Amberlite™ IRA-402, Cl-form, ion-exchange resin, Thermo Scientific Chemicals
Amberlite™ IRA-402, Cl-form, ion-exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amberlite™ IRA-402, Cl-form, ion-exchange resin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 369942500
Price (USD)/ Each
37.65
Online exclusive
44.00 
Save 6.35 (14%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
44.00 
Save 6.35 (14%)
Add to cart
Amberlite™ IRA-402, Cl-form, ion-exchange resin, Thermo Scientific Chemicals
Catalog number369942500
Price (USD)/ Each
37.65
Online exclusive
44.00 
Save 6.35 (14%)
-
Add to cart
Chemical Identifiers
MDL NumberMFCD00145564
CAS52439-77-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Total volume capacity>=1.20 eq/l (Cl)
Harmonic mean size0.600 to 0.750 mm
Moisture holding capacity49.0 to 60.0 % (Cl)
Uniformity coefficient=<1.60
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only