2,4-Dimethoxybenzylamine, 98%, Thermo Scientific Chemicals
2,4-Dimethoxybenzylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dimethoxybenzylamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 369950250
Price (USD)/ Each
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-(2,4-dimethoxyphenyl)methanamine
Molecular FormulaC9H13NO2
InChI KeyQOWBXWFYRXSBAS-UHFFFAOYSA-N
SMILESCOC1=CC=C(CN)C(OC)=C1
Molecular Weight (g/mol)167.21
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5480 to 1.5500 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only