Sodium selenate, 98%, Thermo Scientific Chemicals
Sodium selenate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium selenate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 369961000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13410-01-0
SpecificationsSpecification SheetSpecification Sheet
Identificationconfirmed
Appearance (Form)Crystalline powder
Assay>=97.5 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only