Thermo Scientific Chemicals

Sodium selenate, 98%, Thermo Scientific Chemicals

Catalog number: 369965000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium selenate, 98%, Thermo Scientific Chemicals

Catalog number: 369965000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 369965000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13410-01-0
Identification
confirmed
Appearance (Form)
Crystalline powder
Assay
>=97.5 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text