Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Description | Color | Phase | Thickness (Metric) |
---|---|---|---|---|
36SP06E3-SM | SMART SPME Fiber: DVB/PDMS (Divinylbenzene/Polydimethylsiloxane), 65 μm, Violet, 3/Pk. | Violet | DVB/PDMS (Divinylbenzene/Polydimethylsiloxane), 65 μm | 65 μm |
36SP01P1-SM | SMART SPME Fiber: PDMS (Polydimethylsiloxane), 7 μm, Green, 1/Pk. | Green | PDMS (Polydimethylsiloxane), 7 μm | 7 μm |
36SP01P3-SM | SMART SPME Fiber: PDMS (Polydimethylsiloxane), 7 μm, Green, 3/Pk. | Green | PDMS (Polydimethylsiloxane), 7 μm | 7 μm |
36SP03P1-SM | SMART SPME Fiber: PDMS (Polydimethylsiloxane), 30 μm, Golden, 1/Pk. | Golden | PDMS (Polydimethylsiloxane), 30 μm | 30 μm |
36SP03P3-SM | SMART SPME Fiber: PDMS (Polydimethylsiloxane), 30 μm, Golden, 3/Pk. | Golden | PDMS (Polydimethylsiloxane), 30 μm | 30 μm |
36SP05T1-SM | SMART SPME Fiber: DVB/Carbon WR/PDMS SMART SPME Fiber, 50/30 μm, Dark Gray, 1/Pk. | Dark Gray | DVB/Carbon WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane), 50/30 μm | 50/30 μm |
36SP05T3-SM | SMART SPME Fiber: DVB/Carbon WR/PDMS SMART SPME Fiber, 50/30 μm, Dark Gray, 3/Pk. | Dark Gray | DVB/Carbon WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane), 50/30 μm | 50/30 μm |
36SP06E1-SM | SMART SPME Fiber: DVB/PDMS (Divinylbenzene/Polydimethylsiloxane), 65 μm, Violet, 1/Pk. | Violet | DVB/PDMS (Divinylbenzene/Polydimethylsiloxane), 65 μm | 65 μm |
36SP08A1-SM | SMART SPME Fiber: Acrylate (Polyacrylate), 85 μm, Gray, 1/Pk. | Gray | Acrylate (Polyacrylate), 85 μm | 85 μm |
36SP08A3-SM | SMART SPME Fiber: Acrylate (Polyacrylate), 85 μm, Gray, 3/Pk. | Gray | Acrylate (Polyacrylate), 85 μm | 85 μm |
36SP08M5-SM | SMART SPME Fibers: Collection of Five Different SMART SPME Fibers | Red/Violet/Dark Blue/Grey/Dark Grey | PDMS Thickness: 100 μmDVB/PDMS Thickness: 65 μmCarbon WR/PDMS Thickness: 95 μmAcrylate Thickness: 85 μmDVB/Carbon WR/PDMS Thickness: 50/30 μm | Various |
36SP09C1-SM | SMART SPME Fiber: Carbon WR/PDMS (Carbon Wide Range), 95 μm, Dark Blue, 1/Pk. | Dark Blue | Carbon WR/PDMS (Carbon Wide Range/ Polydimethylsiloxane), 95 μm | 95 μm |
36SP09C3-SM | SMART SPME Fiber: Carbon WR/PDMS (Carbon Wide Range) , 95 μm, Dark Blue, 3/Pk. | Dark Blue | Carbon WR/PDMS (Carbon Wide Range/ Polydimethylsiloxane), 95 μm | 95 μm |
36SP10P1-SM | SMART SPME Fiber: PDMS (Polydimethylsiloxane), 100 μm, Red, 1/Pk. | Red | PDMS (Polydimethylsiloxane), 100 μm | 100 μm |
36SP10P3-SM | SMART SPME Fiber: PDMS (Polydimethylsiloxane), 100 μm, Red, 3/Pk. | Red | PDMS (Polydimethylsiloxane), 100 μm | 100 μm |
Automate solvent-free solid phase micro extraction (SPME) sample preparation by performing sample extraction and enrichment in a single step with our SMART SPME Fibers.
SMART SPME Fibers also offer the additional benefit of tracking usage information. SMART SPME Fibers are compatible with all TriPlus™ RSH instruments as well as PAL platforms and AOC-6000 series instruments.
SMART Technology Benefits