Thermo Scientific Chemicals

2-Ethoxyphenol, 98%, Thermo Scientific Chemicals

Catalog number: 370035000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Ethoxyphenol, 98%, Thermo Scientific Chemicals

Catalog number: 370035000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 370035000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-71-3
IUPAC Name
2-ethoxyphenol
Molecular Formula
C8H10O2
InChI Key
MOEFFSWKSMRFRQ-UHFFFAOYSA-N
SMILES
CCOC1=CC=CC=C1O
GC
>=97.5 %
Appearance (Color)
Clear colorless to yellow to pink
Appearance (Form)
Liquid or solid
Infrared spectrum
Conforms
Refractive index
1.5270 to 1.5310 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text