Thermo Scientific Chemicals

(±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, 95%, Thermo Scientific Chemicals

Catalog number: 370040050
Each
Thermo Scientific Chemicals

(±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, 95%, Thermo Scientific Chemicals

Catalog number: 370040050
Each
Catalog number: 370040050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
88568-95-0
IUPAC Name
methyl (2S)-2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
Molecular Formula
C13H18NO7P
InChI Key
GSYSFVSGPABNNL-NSHDSACASA-N
SMILES
COC(=O)[C@@H](NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
Appearance (Color)
White
Infrared spectrum
Conforms
Melting point
77°C to 80°C
HPLC
>=94 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text