Acetoxyacetyl chloride, 97%
Acetoxyacetyl chloride, 97%
Acetoxyacetyl chloride, 97%
Acetoxyacetyl chloride, 97%
Thermo Scientific Chemicals

Acetoxyacetyl chloride, 97%

CAS: 13831-31-7 | C4H5ClO3 | 136.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 370070050
Price (USD)
35.30
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
35.30
Each
Add to cart
Chemical Identifiers
CAS13831-31-7
IUPAC Name2-chloro-2-oxoethyl acetate
Molecular FormulaC4H5ClO3
InChI KeyHZDNNJABYXNPPV-UHFFFAOYSA-N
SMILESCC(=O)OCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4255 to 1.4275 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link