Zinc acetate, 99%, pure, Thermo Scientific Chemicals
Zinc acetate, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc acetate, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 370081000
Price (USD)/ Each
253.65
Online exclusive
282.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
253.65
Online exclusive
282.00 
Save 28.35 (10%)
Add to cart
Zinc acetate, 99%, pure, Thermo Scientific Chemicals
Catalog number370081000
Price (USD)/ Each
253.65
Online exclusive
282.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS557-34-6
IUPAC Namezinc(2+) diacetate
Molecular FormulaC4H6O4Zn
InChI KeyDJWUNCQRNNEAKC-UHFFFAOYSA-L
SMILES[Zn++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White
Infrared spectrumConforms
Titration Complexometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only