Thermo Scientific Chemicals

2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals

Catalog number: 370110050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals

Catalog number: 370110050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 370110050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
31106-82-8
IUPAC Name
2-(bromomethyl)pyridine
Molecular Formula
C6H6BrN
InChI Key
OFPWMRMIFDHXFE-UHFFFAOYSA-N
SMILES
BrCC1=CC=CC=N1
Appearance (Color)
White to pink to orange-brown
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Titration with HClO4
97.5 to 102.5 %
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text